Orders LinkContactUsLinkComponent
VWR® Contour™ Kemstrut Assemblies
VWR® Contour™ Kemstrut Assemblies
VWR® Contour™ Kemstrut Assemblies
VWR® Contour™ Kemstrut Assemblies
Adjustable-height floor-mounted support for the rear overhang of fume hood worktops.
  • Smooth, easy-to-maintain surfaces
  • Color: Light neutral
It can also be used as a worktop and/or a drainline and piping support in the plumbing space of 363/4x31" wall tables and 363/4x54" or 363/4x55" center tables.
Product Family Options
  • Description
    Material
    Width
    Depth
    Height
  • Support
    Steel
    24½"
    8"
    33 - 37"
    Each (1UN)
    Each (1UN) - $2,051.27
    $2,051.27
    Each (1UN)
       $2,051.27
  • Support 36x14
    Steel
    14"
    33 - 37"
    Each (1UN)
    Each (1UN) - $2,125.70
    $2,125.70
    Each (1UN)
       $2,125.70