KoolTemp® Insulated Shippers, EPS Foam Shipping Coolers, Cold Chain Technologies
Proveedor: COLD CHAIN TECHNOLOGIES
Certificados
Acerca de este artículo
Lightweight and reusable insulated expand polystyrene (Styrofoam) shippers provide excellent thermal protection for perishables and samples.
- Reusable
- Excellent thermal protection
Shippers are available with or without outer corrugated carton.
Entrega: Shippers are available with or without outer corrugated carton.
Frequently Bought Together