Orders LinkContactUsLinkComponent
VWR® Replica-Plating Tool
VWR® Replica-Plating Tool
VWR® Replica-Plating Tool
Supplier:  VWR International
Certificates
VWR® Replica-Plating Tool
VWR® Replica-Plating Tool
2 Options Available Below
Product Details & Documents
The replica-plating tool utilizes a locking ring which secures sterile velveteen squares onto a PVC cylinder for easy control of the plating device.
  • For use with 90 to 100 mm Petri dishes
  • May be disinfected between uses with a brief rinse in 70% ethanol or chlorine bleach
  • Aluminum ring is 10.2 cm (4") diameter; PVC cylinder is 6.9 cm (23/4") high
  • Not autoclavable
To produce replicas, the Petri dish carrying microbial colonies is inverted and lowered onto the cloth surface, then lifted. A fresh sterile nutrient plate is then lowered onto the velveteen surface and lifted, leaving a replica of the original colonies in the new dish.
 : Tool comes with 12 sterile, white, 100% cotton, absorbent twill-backed velveteen squares; 15.2×15.2 cm (6×6"). Replacement velveteen squares are available; packed in a sealed bag with three inner locking bags each containing 12 squares, 36 pcs. total.
Documents
Product Family Options
Información del producto
  • Availability
    Price
  • 25395-380
    Replica-Plating Tool
    Replica-Plating Tool
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $3,953.53
    $3,953.53
    Each (1UN)
       $3,953.53
  • 89033-116
    Velveteen Squares, Black
    Velveteen Squares, Black
    Log in Para disponibilidad
    Log in Para disponibilidad
    Paquete de 36
    Paquete de 36 - $4,590.12
    $4,590.12
    Paquete de 36
       $4,590.12