ContactUsLinkComponent
Analyslide® Petri Dish, Cytiva (Formerly Pall Lab)
Analyslide® Petri Dish, Cytiva (Formerly Pall Lab)
  28145-473
 :  Cytiva
Product available on GSA Advantage®
undefined
Analyslide® Petri Dish, Cytiva (Formerly Pall Lab)
  28145-473
 :  Cytiva
 :  7231
Product available on GSA Advantage®
  • Descripción:
    Petri Dish on a Slide
  • Tamaño:
    47 mm
  • Referencia:
    28145-473
  • Nº de proveedor:
    7231
The Analyslide® Petri dish has a rectangular base and a circular chamber that accepts a 47 mm membrane filter. The sample is protected during microscopic examination, and membrane filters can be stored and protected for reference.
  • Rectangular base fits most microscope stages
  • Inner cover ring for secure closure
  • Clear lid does not interfere with viewing sample
  • Frosted area on base permits identification of the sample with marker
  • Convenient box simplifies collection and storage of samples
Base and lid are constructed from PS.