ContactUsLinkComponent
RODAC™ Plate, Specialty Petri Dish, Sterile, BD Biosciences
  90000-004
undefined
RODAC™ Plate, Specialty Petri Dish, Sterile, BD Biosciences
  90000-004
 :  210340
  • Descripción:
    RODAC® Plate
  • Referencia:
    90000-004
  • Nº de proveedor:
    210340
RODAC® (Replicate Organism Detection and Counting) was developed for environmental studies to determine levels of microbial contamination of surfaces
When plate is filled with 12–13mL of media, the media surface is raised to a convex shape. Area of agar bed is approximately 25cm2 with Quebec-style grids of one centimeter squares. A cake-box type lid protects the agar. Polystyrene, with cover. Packaged 20 per sleeve.