ContactUsLinkComponent
Falcon® Petri Dishes, 50×9 mm, Tight-Fit Lid, Sterile, Corning
Falcon® Petri Dishes, 50×9 mm, Tight-Fit Lid, Sterile, Corning
Catálogo # 25369-022
Proveedor:  Corning
undefined
Falcon® Petri Dishes, 50×9 mm, Tight-Fit Lid, Sterile, Corning
Catálogo # 25369-022
Proveedor:  Corning
Número de proveedor:  351006
Especificaciones
  • Descripción:
    Falcon* Petri Dishes
  • Tamaño:
    50 x 9 mm
  • Referencia:
    25369-022
  • Nº de proveedor:
    351006
Especificaciones
Acerca de este artículo
PS. Manufactured in accordance with the current FDA Quality System.
  • Flat, distortion free optics
  • Stacking rings allow for easier stacking and handling
  • Durable construction for stable dish manipulation
Small diameter saves media and space. Easily sealed with tape for collecting and transporting specimens. Polystyrene. Sterilized by gamma irradiation. Packed 20 per bag.