Orders LinkContactUsLinkComponent
Electrolyte Solution
Electrolyte Solution
Electrolyte 1 M LiCl/EtOH, 6×30 ml, Bottle, pH, —
Catalog # 76359-846
Supplier #:  51350088
Electrolyte Solution
Catalog # 76359-846
CAS Number:  
Electrolyte Solution
Electrolyte Solution
Electrolyte 1 M LiCl/EtOH, 6×30 ml, Bottle, pH, —
CAS Number:  
Price
$8,284.14
Log in for availability
Paquete de 6
Product Details & Documents
pH electrolyte and maintenance solution.
Documents
Specifications
Specifications
  • Pack type:
    Bottle
  • Description:
    Electrolyte 1 M LiCl/EtOH
  • Size:
    6×30 ml
  • For:
    pH
  • Ion:
Specifications