Nunclon™ Delta MultiDishes, Sterile, Thermo Scientific
73521-006
- Nº de pocillos:24
- Referencia:73521-006
- Empaquetado:5/Sleeve
- External Dimensions:128×86 mm
- Well Area:1.9 cm2
- Working Well Volume:0.4 - 1.0 ml
- Nº de proveedor:142485
These polystyrene dishes with flat-bottom wells incorporate the Nunclon™Δ treated surface for optimal cell attachment and growth
Lids are keyed for a one-way fit. Rings in the lids reduce evaporation and condensation. Writing areas on both lids and dishes and individual alphanumeric markings allow easy identification of samples. Nontoxic and certified nonpyrogenic. Stackable.