Barnstead/Lab-Line Clamps for Universal Platforms, Thermo Scientific
Proveedor: THERMO FISHER SCIENTIFIC MX
Certificados
Acerca de este artículo
Use with Thermo Scientific universal platforms (57018-990 series)
Clamp 57019-692 accommodates a 2.5 l low-form culture flask. Clamp 57019-696 accommodates a 2.8 l Fernbach flask. Clamp 47742-748 accommodates a microplate or deep well plate. All other clamps are for Erlenmeyer flasks.
Clamps are constructed of stainless steel with retaining springs to hold vessels securely in place.
Información relacionada
Thermo Scientific MaxQ-Shakers Thermo Scientific Solaris Orbital Shakers Thermo Scientific Solaris Shaker
Documentación del producto
Frequently Bought Together