ContactUsLinkComponent
Nunc™ Cell Culture/Petri Dishes, Thermo Scientific
Catalog # 25382-334
undefined
Nunc™ Cell Culture/Petri Dishes, Thermo Scientific
Catalog # 25382-334
Supplier Number:  153066
Specifications
  • Ø:
    35 mm
  • Descripción:
    Nunclon™Δ Treated
  • Altura:
    10 mm
  • Referencia:
    25382-334
  • Growth Area:
    8.8 cm²
  • Nº de proveedor:
    153066
Specifications
About this item
Perform a variety of applications across a broad range of sizes and styles to suit a variety of experimental needs with Thermo Scientific™ Nunc™ Cell Culture/Petri Dishes. These dishes receive the Thermo Scientific™ Nunclon™ Delta surface treatment, a proprietary surface modification that promotes maximum adhesion for a broad range of cell types.
These cell culture/petri dishes are made of polystyrene.
These dishes receive the Thermo Scientific™ Nunclon™ Delta surface treatment, a proprietary surface modification that promotes maximum adhesion for a broad range of cell types.
Ordering information: Quality certificates are included in each case.