Orders LinkContactUsLinkComponent
Corning® Microcentrifuge Tubes, Polypropylene
Corning® Microcentrifuge Tubes, Polypropylene
Supplier:   Corning
Corning® Microcentrifuge Tubes, Polypropylene
Supplier:  Corning
Certificates
Corning® Microcentrifuge Tubes, Polypropylene
Corning® Microcentrifuge Tubes, Polypropylene
4 Options Available Below
Product Details & Documents
Ideal for convenient handling and recovery of small samples. External graduations and frosted writing surface aid in sample identification. Tubes are provided with flat snap caps. Polypropylene tubes available in natural or rainbow colors. Certified RNase- and DNase-free. Nonsterile. Autoclavable.
  • External graduations and frosted writing spot for easy sample identification
  • Positive seal design allows for repeated opening and closing
  • Flat cap surface for convenient labeling
  • Withstands a maximum of 12320 rpm
These microcentrifuge tubes are made of natural (uncoloured) or colored polypropylene and are certified RNase-/DNase-free.
Assorted color pack includes one bag each of blue, green, yellow, red and orange tubes.
Ordering information: Optional autoclavable plastic tube racks sold separately.
Product Family Options
Product Information
  • Volume
    Color
    Maximum RCF
    Packaging
    Availability
    Price
  • Tube
    0.65 ml
    Clear
    17000×g
    500/bag
    Log in for availability
    Log in for availability
    Case (1,000UN)
    Case (1,000UN) - $3,270.00
    $3,270.00
    Case (1,000UN)
       $3,270.00
    Supplier Number: 3208
  • Tube
    0.65 ml
    Rainbow Pack
    17000×g
    200/bag
    Log in for availability
    Log in for availability
    Case (1,000UN)
    Case (1,000UN) - $3,270.00
    $3,270.00
    Case (1,000UN)
       $3,270.00
    Supplier Number: 3209
  • Tube
    1.7 ml
    Clear
    17000×g
    500/bag
    Log in for availability
    Log in for availability
    Case (500UN)
    Case (500UN) - $1,370.00
    $1,370.00
    Case (500UN)
       $1,370.00
    Supplier Number: 3620
  • Tube
    2 ml
    Clear
    17000×g
    500/bag
    Log in for availability
    Log in for availability
    Case (1,000UN)
    Case (1,000UN) - $2,850.00
    $2,850.00
    Case (1,000UN)
       $2,850.00
    Supplier Number: 3213