PYREX® VISTA™ Test Tubes, Reusable, Corning®
Catálogo # 89090-340
Proveedor: Corning
Product available on GSA Advantage®
Especificaciones
- Volumen:20 ml
- Environmentally Preferable:Low Manufacturing Impact
- Certificación ambiental:I-RECs, ISO 14001 - Monterrey
- Referencia:89090-340
- Tube Bottom Shape:5
- O.D.×L:16×150 mm
- Nº de proveedor:70800-16
Especificaciones
Acerca de este artículo
Annealed test tubes offer excellent heat resistance and chemical stability. Reusable tubes have a round-bottom design and feature fire-polished rims. All tubes except the 55 and 70mL sizes have a blue enamel marking spot. Supplied without caps. Nonsterile.
- Non-sterile
Reusable tubes have a round-bottom design and feature fire-polished rims. All tubes except the 55 and 70 ml sizes have a blue enamel marking spot.
Entrega: Supplied without caps