Orders LinkContactUsLinkComponent
Electrode Fill Solutions
Electrode Fill Solutions
Electrode Fill Solutions
Certificates
Electrode Fill Solutions
Electrode Fill Solutions
2 Options Available Below
Product Details & Documents
FRISCOLYT-B™ (84.4% glycerol, 11% KCl, and 4.6% H₂O) is for low temperature samples.
Delivery information: Supplied in 25 ml (0.9 oz.) bottles.
Documents
Product Family Options
Product Information
  • Pack type
    Volume
    Ion
    Availability
    Price
  • 1M KNO₃
    Bottle
    25 ml
    Sulfide, cadmium, cyanide, iodide, chloride, thiocyanate, lead, copper, bromide
    Log in for availability
    Log in for availability
    Each (25ml)
    Each (25ml) - $612.35
    $612.35
    Each (25ml)
       $612.35
    Supplier Number: 51343182
  • 97001-714
    3M KCl saturated with AgCl
    3M KCl saturated with AgCl
    Bottle
    25 ml
    Log in for availability
    Log in for availability
    Each (25ml)
    Each (25ml) - $990.29
    $990.29
    Each (25ml)
       $990.29
    Supplier Number: 51343184