PYREX® Separating Funnels, Cylindrical, Glass, Corning
100 ml, 294 mm, 24/40, 4, 5 ml
Supplier: Corning
Supplier #: 6383A-100
PYREX® Separating Funnels, Cylindrical, Glass, Corning
Catalog # 32645-163
Supplier: Corning
Product available on GSA Advantage®
Jump to:
Product Details & Documents
PYREX®, borosilicate glass, with glass stopper.
With socket and plain stem. Available with interchangeable PTFE key for grease-free performance. With graduation.
Documents
Certificates
Buscar por número de loteEspecificaciones
- Graduation Interval:5 ml
- Capacity:100 ml
- Length:294 mm
- ST Joint Size:24/40
- ST Stopcock Size:4
- Cat. No.:32645-163
- Environmental certification:I-RECs, ISO 14001 - Monterrey
- Environmentally Preferable:Low Manufacturing Impact