Coverage Plus NPD® One-Step Cleaner Disinfectants
21899-844
- Tamaño:3.78 L (1 gal.)
- Referencia:21899-844
- Empaquetado:Plastic bottle, 4/case
- Nº de proveedor:638708
STERIS' Coverage Plus NPD® One-Step Cleaner disinfectant is a liquid concentrate specifically formulated for the disinfection of hard surfaces. The use-dilution can be applied manually or sprayed.
- Efficacy - Quaternary ammonium active ingredients provide broad spectrum antimicrobial efficacy
- Productivity - Formulation includes surfactants for surface cleaning while disinfecting
- Ease of Use - Ideal for use in a contamination control rotation
- Reliability - Complete lot traceability
- Assurance - Each lot documented by Certificate of Analysis
STERIS' Coverage Plus NPD® One-Step Cleaner disinfectant a phosphate-free, one-step cleaner/disinfectant formulated for use on hard non-porous surfaces. This product consists of four quaternary ammonium compounds formulated with a compatible detergent system. It is proven effective in the presence of heavy soil (5% bovine serum) and hard water. Coverage Plus NPD® disinfectant contains no perfumes or dyes that could interfere with critical research studies or manufacturing processes. Use-dilution is 1:256.
: Danger, GHS02; GHS05; GHS07.