PYREX® Test Tubes, Reusable, Borosilicate Glass, Corning®
: Corning
Made from special machine-made tubing of correct wall weight. Tubes are well annealed, resistant to heat, and chemically stable. Rims are fire polished. Particularly suited for use in bacteriological work. Repeated sterilization, wet or dry, does not cause clouding.
They are well annealed, resistant to heat, and chemically stable. Rims are fire-polished.
- Catalog No:
- 60815-228
- 60815-240
- 60815-068
- 60815-104
- 60815-126
- 60815-160
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Certificación ambiental:
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
Frequently Bought Together