Orders LinkContactUsLinkComponent
KIMAX® Test Tubes, Plain, Kimble Chase
55 ml, 150 mm, 25 mm
  89001-450
 :  45050-25150
KIMAX® Test Tubes, Plain, Kimble Chase
  89001-450
KIMAX® Test Tubes, Plain, Kimble Chase
KIMAX® Test Tubes, Plain, Kimble Chase
55 ml, 150 mm, 25 mm
$2,330.05
Paquete de 72 Case de 4 (72UN)
KIMAX-51® tubes are constructed from N-51A borosilicate glass tubing and feature a round bottom, sturdy lip, and uniform wall thickness for maximum heat transfer and chemical resistance.
Tubes are supplied without a marking spot or cap.
  • Ø:
    25 mm
  • Capacity:
    55 ml
  • Height:
    150 mm