CELLSTAR® Centrifuge Tubes, Polypropylene, Sterile, Greiner Bio-One
Proveedor: GREINER BIO-ONE
Certificados
Acerca de este artículo
These conical-bottom, sterile centrifuge tubes are made from ClearChoice™ polypropylene with excellent optical clarity and high RCF ratings.
- Sterile
- Free of RNase, DNase, and human DNA
- Non-pyrogenic
- Available with or without a skirt
- Polypropylene construction
Tubes feature graduations and an ID field printed on the side.
Ordering information: Supplied with HDPE flat-top screw caps.
Información relacionada
Certificados
Buscar por número de lote82050-280
Additional Product Details
AM-188261 AM-188261-1 AM-188271 AM-188271-1 AM-188281
Datasheet
More
Greiner Bio-One Product Catalog
Product Literature
Frequently Bought Together