Axygen® Conical-Bottom Centrifuge Tubes, Polypropylene, Corning
Supplier: Corning
Certificates
About this item
Polypropylene tubes feature printed graduations and a writing area for sample identification.
- Caps may be closed with one hand
- Tubes are autoclavable
The 15 ml tubes can be centrifuged to 7500 g; the 50 ml tubes can be centrifuged to 9400 g.
Specifications
- Catalog No:
- 89005-588
- 89005-624
- 89005-592
- 89005-622
- 89005-586
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Certificación ambiental:
- I-RECs, ISO 14001 - Reynosa
- I-RECs, ISO 14001 - Reynosa
- I-RECs, ISO 14001 - Reynosa
- I-RECs, ISO 14001 - Reynosa
- I-RECs, ISO 14001 - Reynosa