KIMAX® Centrifuge Tubes, Graduated, Conical-Bottom, with Screw Caps, Kimble Chase
Proveedor: DWK LIFE SCIENCES S. DE R.L. DE MX
Certificados
Acerca de este artículo
Tubes are graduated and calibrated to contain.
- Centrifuge tubes are made of borosilicate glass
Scale, legend, and marking spot are printed in durable white ceramic enamel. Tubes have a screw-thread finish and include an autoclavable phenolic cap with PTFE-faced rubber liner. Heavy-duty tubes are made with heavier walled tubing to withstand higher centrifugation speeds.
Frequently Bought Together