Nalgene® Round-Bottom Centrifuge Tubes, Polypropylene Copolymer, Thermo Scientific
Catalog # 21010-749
Supplier: Thermo Fisher Scientific
Specifications
- Ø×L:15.9×103 mm
- Volumen:12 ml
- Descripción:Round-Bottom Tube
- Referencia:21010-749
- RCF máximo:50000×g
- Fits:Closure No. 16
- Nº de proveedor:3110-0120
Specifications
About this item
Heavy-duty construction for repeated autoclavability.
- Excellent chemical and abrasion resistance
- An economical choice for refrigerated or standard centrifugation
Note: For proper performance, fill tubes to at least 80% of the total capacity.
Molded for uniform wall thickness and extra strength. PPCO has a better low-temperature rating than standard polypropylene. Also useful as test tubes. RCF ratings are based on chemical compatibility and proper rotor fit.
Ordering information: Supplied without closures. Polypropylene closures available separately (see 21010-965 series).