Nalgene® Round-Bottom Centrifuge Tubes, Polycarbonate, Thermo Scientific
21009-160
- Ø×L:16×103.5 mm
- Volumen:12 ml
- Descripción:Round-Bottom Tube
- Referencia:21009-160
- RCF máximo:50000×g
- Fits:Closure No. 16
- Nº de proveedor:3117-0120
Strong, crystal clear polycarbonate tubes for use with temperatures from –135 to 135 °C (–211 to 275 °F).
- For use in refrigerated and non-refrigerated rotors
- Non-toxic
- Autoclavable
Note: For proper performance, fill tubes to at least 80% of the total capacity.
Special molding technique provides uniform wall thickness for greater strength.
: Supplied without closures. Polypropylene closures available separately (see 21010-852 series).