Orders LinkContactUsLinkComponent
pH Buffer Capsules, Hydrion®
pH Buffer Capsules, Hydrion®
pH Buffer Capsules, Hydrion®
Certificates
pH Buffer Capsules, Hydrion®
pH Buffer Capsules, Hydrion®
3 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
For the quick and convenient preparation of fresh solutions of known pH values
Capsule contents dissolve in 100mL of distilled water, giving a clear solution accurate to ±0.02 pH unit. The dry powder remains stable. Each vial contains 10 capsules.
Documents
Product Family Options
Product Information
  • pH Accuracy
    pH
    Color
    Availability
    Price
  • ±0.02
    4.00
    Clear
    Log in for availability
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $283.25
    $283.25
    Log in for availability
    Case de 5 (1UN)
    Case de 5 (1UN) - $308.74
    $308.74
    Each (1UN)
       $283.25
    Case de 5 (1UN)
       $308.74
    Supplier Number: 270-4.00
  • ±0.02
    7.00
    Clear
    Log in for availability
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $283.25
    $283.25
    Log in for availability
    Case de 5 (1UN)
    Case de 5 (1UN) - $308.74
    $308.74
    Each (1UN)
       $283.25
    Case de 5 (1UN)
       $308.74
    Supplier Number: 270-7.00
  • ±0.02
    10.00
    Clear
    Log in for availability
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $283.25
    $283.25
    Log in for availability
    Case de 5 (1UN)
    Case de 5 (1UN) - $308.74
    $308.74
    Each (1UN)
       $283.25
    Case de 5 (1UN)
       $308.74
    Supplier Number: 270-10.00
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 