VWR® Centrifuge Tubes, Polystyrene
Proveedor: VWR International
Certificados
Acerca de este artículo
These tubes are made of optically clear PS and feature distinct molded graduations.
- Colorless and transparent are perfect for biological reaction experiments
- RNase-, DNase-, and Endotoxin-Free
- Leak-proof
- Not autoclavable
- Sterile tubes are sterilized by irradiation
- During the freezing process, it is better to use plastic rack
- During high-speed centrifugation, it is better to use suitable rotor adapters
The flat surface of the polypropylene cap provides an at-a-glance identification area.
Certificaciones: Manufactured in a class 100000 room environment. Manufactured under ISO13485:2016 and ISO9001:2015 quality management. Meet the requirements of RoHS.
Información de pedido: Choose from tubes and caps supplied in Earth Friendly cardboard racks, bag packed, or bulk packed.
Especificaciones
- Catalog No:
- 21008-197
- 21008-189
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Certificación ambiental:
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
Especificaciones
Información relacionada
Frequently Bought Together