Safety Face Shield, Sellstrom®
Headgear with Window, 1 mm (0.04")
Catalog # 33007-060
Supplier: SUREWERX USA INC
Supplier #: S39010
Jump to:
Product Details & Documents
Provides facial protection from splashes and flying particles.
- Economical, lightweight design
- one-piece, molded headgear with free-floating suspension
- Two snap-in locking headgear
- Straps adjust independently to ensure a tight fit
Replaceable clear acetate window: 1 mm (0.04") thick
WxH: 8x12"
Caution: Note: Face shield should be worn over ANSI-approved protective eyewear.
Specifications
- Description:Headgear with Window
- Thickness:1 mm (0.04")