ContactUsLinkComponent
VWR® Slide Mailers for 5 Slides
VWR® Slide Mailers for 5 Slides
Catálogo # 82024-598
Proveedor:  VWR International
undefined
VWR® Slide Mailers for 5 Slides
Catálogo # 82024-598
Proveedor:  VWR International
Especificaciones
  • Color:
    Natural
  • Descripción:
    Side-Opening Mailer
  • Referencia:
    82024-598
  • An x Prf x Al:
    81 x 19 x 31 mm (33/16 x 3/4 x 17/32")
Especificaciones
Acerca de este artículo
Ideal for shipping, storing, or staining slides, these polypropylene containers hold five standard 25x75 mm (1x3") slides.
Inside wall features slotted channels to keep slides separated and make removal easier. Closes securely to keep slides safe.
Información de pedido: Assorted colors include red, yellow, blue, purple, and orange.