Preparative TLC Plates, Analtech
Silica gel, 200×200 mm, 2000 µm, 60 Å, Calcium sulfate
Catalog # 80086-352
Small Business Enterprise
Supplier: MILES SCIENTIFIC CORP
Supplier #: P02015
Preparative TLC Plates, Analtech
Catalog # 80086-352
Supplier: MILES SCIENTIFIC CORP
Small Business Enterprise
Jump to:
Product Details & Documents
Preparative TLC plates feature thicker adsorbent layers and are useful for separation of larger quantities of material than plates with a 250 µm thick coating.
- Thicker layers for higher capacity
- Full range of plate sizes
- Prep-scored option for convenient zone location
- Available with preadsorbent sample loading zone
- Calcium sulfate binder soft later for easy scraping
When materials to be separated are applied in long streaks, specific compounds can be recovered after development by scraping the adsorbent layers from the plate in the region of interest and eluting the separated material from the adsorbent using a strong solvent.
Specifications
- Coating material:Silica gel
- Layer thickness:2000 µm
- Plate size:200×200 mm
- Pore size:60 Å
- Binder:Calcium sulfate
- Fluorescent Indicator:No
- Cat. No.:80086-352
- Average particle size:10 µm