TLC Developing Chamber, Cylindrical, Chemglass
Supplier: CHEMGLASS INC.
Jump to:
Acerca de este artículo
Cylindrical glass chamber for development of TLC plates from 25x75mm to 50x100mm
Chamber has a short length inner with a tool ball style handle. Chambers 80066-462 and 80066-464 have a hex base for added stability. Complete chamber is supplied with bottle and stopper.
1 Options Available Below
Product Details & Documents
Cylindrical glass chamber for development of TLC plates from 25x75mm to 50x100mm
Chamber has a short length inner with a tool ball style handle. Chambers 80066-462 and 80066-464 have a hex base for added stability. Complete chamber is supplied with bottle and stopper.
Product Family Options
Información del producto
- For Use WithAvailabilityPrice
- 80066-464TLC developing chamber, 34/15 joint sizeTLC developing chamber, 34/15 joint size25×100 mm platesDiscontinued-Supplier Number: CG-1181-02
- 80066-466TLC developing chamber, 60/15 joint sizeTLC developing chamber, 60/15 joint size50×100 mm platesLog in Para disponibilidadEach (1UN)$6,190.00Supplier Number: CG-1181-03