Orders LinkContactUsLinkComponent
BD Vacutainer® Eclipse™ Blood Collection Needles, BD Diagnostics
BD Vacutainer® Eclipse™ Blood Collection Needles, BD Diagnostics
Needle, 3.175 cm (11/4"), 22 G
Catalog # VT-368608
Número de proveedor #:  368608
BD Vacutainer® Eclipse™ Blood Collection Needles, BD Diagnostics
Catalog # VT-368608
BD Vacutainer® Eclipse™ Blood Collection Needles, BD Diagnostics
BD Vacutainer® Eclipse™ Blood Collection Needles, BD Diagnostics
Needle, 3.175 cm (11/4"), 22 G
Price
$232.00
Log in Para disponibilidad
Paquete de 48 Case de 10 (48UN)
Product Details & Documents
These sterile, safety-engineered, multi-sample blood collection needles offer a simple, effective way to collect blood while reducing the possibility of needlestick injuries. They feature a patented safety shield that allows for one-handed activation to cover the needle completely upon withdrawal from the vein, and confirm proper activation with an audible click. The safety shield is already aligned with the bevel of the needle, so no extra assembly or adjustment is required by the user. The intuitive design of the advanced protection technology means there is no need to change the standard, one-handed venipuncture technique when using these needles. Needles with pre-attached holders help facilities remain single-use holder compliant.
They feature a patented safety shield that allows for one-handed activation to cover the needle completely upon withdrawal from the vein, and confirm proper activation with an audible click. The safety shield is already aligned with the bevel of the needle, so no extra assembly or adjustment is required by the user. The intuitive design of the advanced protection technology means there is no need to change the standard, one-handed venipuncture technique when using these needles. Needles with pre-attached holders help facilities remain single-use holder compliant.
Especificaciones
  • Description:
    Needle
  • Needle Length:
    3.175 cm (11/4")
  • Needle Gauge:
    22 G