ContactUsLinkComponent
SP Bel-Art ProCulture Round Microcentrifuge Floating Bubble Racks, Bel-Art Products, a part of SP
  60986-342
undefined
SP Bel-Art ProCulture Round Microcentrifuge Floating Bubble Racks, Bel-Art Products, a part of SP
  60986-342
 :  F18875-1000
  • Ø orificio:
    10.8 mm (⁷/₁₆")
  • Núm. de orificios:
    20
  • Descripción:
    Bubble racks for 1000 ml beakers, O.D. 95 mm (3¾")
  • Referencia:
    60986-342
  • For Tubes:
    1.5 ml
  • Nº de proveedor:
    188751000
Made especially for use with beakers and waterbaths, these space-saving racks are ideal for small quantities of tubes and for use in cell culture laboratories.
  • Buoyant TPX® brand polymethylpentene with tiny bubbles throughout for additional buoyancy
  • Insert tubes from the top and easily remove them by pressing the rack down on the bench
  • Withstands boiling for 30 minutes at 100 °C (212 °F)
  • Steam autoclavable at 121 °C (250 °F)
  • Leg length: 19 mm (3/4"); thickness: 6.4 mm (1/4")
Perfect for incubating microtubes in a beaker of water.