Orders LinkContactUsLinkComponent
VWR® Microscope Slide Boxes for 25 Slides
VWR® Microscope Slide Boxes for 25 Slides
VWR® Microscope Slide Boxes for 25 Slides
Supplier:  VWR International
Certificates
VWR® Microscope Slide Boxes for 25 Slides
VWR® Microscope Slide Boxes for 25 Slides
3 Options Available Below
Product Details & Documents
These heavy-duty, two-piece slide boxes feature cork lining and store up to 25 standard 25x75 mm (1x3") slides.
  • Heavy-duty design
  • Color-coded for easy identification
Boxes stack securely on top of each other. Ideal for storage or transport. Dimensions: 14Lx8.9Wx3.2Dcm (51/2x31/2x11/4").
Product Family Options
Product Information
  • Color
    Availability
    Price
  • Blue
    Log in for availability
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $80.33
    $80.33
    Log in for availability
    Case de 72 (1UN)
    Case de 72 (1UN) - $87.56
    $87.56
    Each (1UN)
       $80.33
    Case de 72 (1UN)
       $87.56
  • Green
    Log in for availability
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $80.33
    $80.33
    Log in for availability
    Case de 72 (1UN)
    Case de 72 (1UN) - $87.56
    $87.56
    Each (1UN)
       $80.33
    Case de 72 (1UN)
       $87.56
  • Red
    Log in for availability
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $80.33
    $80.33
    Log in for availability
    Case de 72 (1UN)
    Case de 72 (1UN) - $87.56
    $87.56
    Each (1UN)
       $80.33
    Case de 72 (1UN)
       $87.56