ContactUsLinkComponent
Slide and Tissue Block Storage Files, Azer Scientific
Slide and Tissue Block Storage Files, Azer Scientific
Catálogo # 95045-512
Proveedor:  AZER SCIENTIFIC INC
Product available on GSA Advantage®
Minority or Woman-Owned Business Enterprise
Slide and Tissue Block Storage Files, Azer Scientific
Catálogo # 95045-512
Proveedor:  AZER SCIENTIFIC INC
Número de proveedor:  ES080
Product available on GSA Advantage®
Minority or Woman-Owned Business Enterprise
Especificaciones
  • Profundidad:
    30.5 cm (12")
  • Anchura:
    28 cm (11")
  • Capacidad:
    500 Blocks
  • Descripción:
    Tissue Block Storage File with 8 Drawers
  • Altura:
    10.2 cm (4")
  • Referencia:
    95045-512
  • Nº de proveedor:
    ES080
Especificaciones
Acerca de este artículo
These storage files provide an economical and efficient solution for storing and indexing microscope slides and tissue blocks.
  • Sturdy and easy to assemble
  • Interlocking Design
  • Capacity of 500 Blocks or 2000 slides
  • Eight-drawer tissue block file
  • Polymer-Coated Surfaces
The stackable, interlocking cardboard construction is easy to assemble.
The four-drawer slide storage file can hold up to 2000 slides. The eight-drawer tissue block file can hold up to 500 blocks. White exterior surfaces are coated with polymer to help resist moisture.