General Frosted and Plain Microscope Slides, Epredia™
Catalog # 48382-180
Proveedor: Erie Scientific
Especificaciones
- Descripción:Plain slides
- l×a:3×2"
- Grosor:1.0 mm
- Referencia:48382-180
- Nº de proveedor:2957F
Especificaciones
Acerca de este artículo
Ideal for use in nonsterile cytology and pathology applications
- Manufactured to the highest standards
- Pre-cleaned for use in hematology, microbiology, urinalysis, and general laboratory applications
- Finely ground edges
- Frosted tab versions for manual labeling or application of printed labels
- Plain versions are completely plain without frosted tab
- Ready to use
Made of water-white, noncorrosive glass for maximum clarity.
Ordering information: Packaged ½ gross per pack (two packs per gross).