Orders LinkContactUsLinkComponent
VWR® Premium Superfrost® Plus Microscope Slides
VWR® Premium Superfrost® Plus Microscope Slides
Superfrost® slides, White, 1.0 mm, 75×25 mm (3×1")
Catalog # 48311-703
Supplier:   VWR International
VWR® Premium Superfrost® Plus Microscope Slides
Catalog # 48311-703
Supplier:  VWR International
VWR® Premium Superfrost® Plus Microscope Slides
VWR® Premium Superfrost® Plus Microscope Slides
Superfrost® slides, White, 1.0 mm, 75×25 mm (3×1")
Price
$865.83
Log in Para disponibilidad
Paquete de 72 Case de 20 (72UN)
Product Details & Documents
The VWR® Superfrost® Plus micro slide features a special treatment that eliminates the need for additional adhesives or protein coatings, and improves tissue adherence in microwave stainings.
  • Ideal for frozen tissue sections and cytology preparation applications
  • Opaque coating reliably retains laboratory markings
  • Positively charged
Slides are specially treated for frozen tissue sections and cytology preparation applications, eliminating the need for special adhesives and protein coatings. Cytology preparations placed on these slides resist cell loss during enzyme digestion, DNA denaturation, and RNA hybridization. Slides also improve tissue adherence during microwave staining. Slides have an opaque coating that retains most laboratory marking methods.
Especificaciones
  • Color:
    White
  • Description:
    Superfrost® slides
  • L×W:
    75×25 mm (3×1")
  • Thickness:
    1.0 mm