VWR® Micro Slides and Coverslips, Plastic
Plastic cover slips, 0.25 mm, 22×22 mm, 10 inner packs of 100
Supplier: VWR International
Jump to:
Product Details & Documents
Efficient, disposable slides are manufactured from optically clear vinyl. Slides will not break, scratch, or cut fingers. Ideal for wet mounts. Micro coverslips are unbreakable and non-curling.
- Made of clear vinyl plastic
- Refractive index as glass
- Unbreakable and safer than glass
- Used in biological and medical labs, as well as educational applications
Slides will not break, scratch, or cut fingers. Ideal for wet mounts. Micro coverslips are unbreakable and non-curling.
Documents
Especificaciones
- Description:Plastic cover slips
- L×W:22×22 mm
- Thickness:0.25 mm
- Packaging:10 inner packs of 100