VWR VistaVision™ Cover Glasses, No. 2
Supplier: VWR International
Certificates
About this item
Microscope cover glasses are made of pure white borosilicate glass with excellent chemical resistance and flatness and are free of bubbles and scratches. Under ultraviolet light, they emit no distrubing self-fluorescence. Suitable for use in coverslippers. Manufactured in accordance with DIN ISO 8255-1.
- Pure white borosilicate glass fabrication
- Excellent chemical resistance and flatness
Under ultraviolet light, they emit no disturbing self-fluorescence. Suitable for use in coverslippers. Manufactured in accordance with DIN ISO 8255-1.
Refractive index: 1.525 ±0.0015 ne
Ordering information: Square and circular cover glasses are supplied 1 oz. per two-part box.
Frequently Bought Together