Orders LinkContactUsLinkComponent
VWR® SuperSlips™ Micro Cover Glasses, Rectangular, No. 1
VWR® SuperSlips™ Micro Cover Glasses, Rectangular, No. 1
VWR® SuperSlips™ Micro Cover Glasses, Rectangular, No. 1
Supplier:  VWR International
Certificates
VWR® SuperSlips™ Micro Cover Glasses, Rectangular, No. 1
VWR® SuperSlips™ Micro Cover Glasses, Rectangular, No. 1
3 Options Available Below
Product Details & Documents
These cover glasses are corrosion-resistant and uniformly thin and flat.
  • Uniformly thin and flat
  • Suitable for humid environments
  • Resistant to corrosion
The processing techniques result in superior cleanliness. Superslip coverslips combine specialized production methods with moisture-resistant packaging to prevent sticking. Suitable for applications in humid environments. Thickness: 0.13 to 0.17 mm. Supplied in 1 oz. increments.
Documents
Product Family Options
Información del producto
  • L×W
    Thickness
    Availability
    Price
  • 48404-457
    Micro cover glasses, rectangular
    Micro cover glasses, rectangular
    40×24 mm
    0.13 - 0.17 mm
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Onza
    Onza - $180.85
    $180.85
    Log in Para disponibilidad
    Case de 10 (1oz)
    Case de 10 (1oz) - $197.13
    $197.13
    Onza
       $180.85
    Case de 10 (1oz)
       $197.13
  • 48404-453
    Cover glasses, rectangular
    Cover glasses, rectangular
    50×24 mm
    0.13 - 0.17 mm
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Onza
    Onza - $154.06
    $154.06
    Log in Para disponibilidad
    Case de 10 (1oz)
    Case de 10 (1oz) - $167.93
    $167.93
    Onza
       $154.06
    Case de 10 (1oz)
       $167.93
  • 48404-455
    Micro cover glasses, rectangular
    Micro cover glasses, rectangular
    60×24 mm
    0.13 - 0.17 mm
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Onza
    Onza - $193.74
    $193.74
    Log in Para disponibilidad
    Case de 10 (1oz)
    Case de 10 (1oz) - $211.17
    $211.17
    Onza
       $193.74
    Case de 10 (1oz)
       $211.17