Orders LinkContactUsLinkComponent
VWR® Micro Cover Glasses, Rectangular, No. 2
VWR® Micro Cover Glasses, Rectangular, No. 2
Cover glasses, rectangular, 50×24 mm, 0.17 - 0.25 mm, #2
  48382-136
VWR® Micro Cover Glasses, Rectangular, No. 2
  48382-136
VWR® Micro Cover Glasses, Rectangular, No. 2
VWR® Micro Cover Glasses, Rectangular, No. 2
Cover glasses, rectangular, 50×24 mm, 0.17 - 0.25 mm, #2
$171.46
Onza Case de 10 (1oz)
Uniformly thin and flat. Processing techniques result in superior cleanliness. No. 2 thickness of 0.17 to 0.25mm.
  • Uniformly thin and flat
  • Processing techniques result in superior cleanliness
No. 2 thickness of 0.17 to 0.25mm.
Documents
  • Numbering:
    #2
  • Description:
    Cover glasses, rectangular
  • L×W:
    50×24 mm
  • Thickness:
    0.17 - 0.25 mm