ContactUsLinkComponent
VWR® Micro Cover Glasses, Round, No. 2
VWR® Micro Cover Glasses, Round, No. 2
  48382-042
undefined
VWR® Micro Cover Glasses, Round, No. 2
  48382-042
  • Ø:
    18 mm
  • Descripción:
    Cover glasses, round
  • Grosor:
    0.17 - 0.25 mm
  • Referencia:
    48382-042
These microscope cover glasses are corrosion-resistant and uniformly thin and flat.
  • Borosilicate glass fabrication
  • Thickness: 0.17-0.25mm
The processing techniques result in superior cleanliness. Manufactured from borosilicate glass, cover glasses are free from bubbles, scratches, and striations.
 : Packaged 1 oz. per plastic box.