VWR® Micro Cover Glasses, Square, No. 11/2
Proveedor: VWR International
Certificados
Acerca de este artículo
These cover glasses offer maximum clarity, superior corrosion resistance, and uniform thickness and size.
- No. 11/2 thickness is 0.16 to 0.19mm
They are free of striae and oily film.
Información de pedido: Packaged 1 oz. per plastic box.
Información relacionada
Frequently Bought Together