VWR® Micro Cover Glasses, Square, No.1
Proveedor: VWR International
Certificados
Acerca de este artículo
VWR® Micro Cover Glasses are square, corrosion-resistant, and uniformly thick and flat.
- Corrosion-resistant
- Uniform thickness
No. 1 thickness is 0.13 to 0.17mm.
Información de pedido: Packaged 1 oz. per plastic box.
Información relacionada
Frequently Bought Together