Sleeve-Style Rubber Stoppers, Wheaton
Mouth with ID×OD 15×30 mm
Supplier: DWK LIFE SCIENCES S. DE R.L. DE MX
Número de proveedor #: 224100-320
Sleeve-Style Rubber Stoppers, Wheaton
Catalog # 16170-462
Supplier: DWK LIFE SCIENCES S. DE R.L. DE MX
Jump to:
Product Details & Documents
Plug stoppers with fold-over skirts are for use on serum vials and other crimp-finish containers that do not require an aluminum seal or overcap
Stoppers may also be used as injection sites for multiple dose entry. General-purpose natural red rubber is excellent for resealing and multi-puncture applications. Diameter: 100mm.
Documents
Especificaciones
- Fits:Mouth with ID×OD 15×30 mm