VWR® One-Hole Kjeldahl Rubber Stoppers, Black
Proveedor: VWR International
Certificados
Acerca de este artículo
These black rubber, one-hole stoppers are specifically designed to fit 500 and 800 mL Kjeldahl nitrogen determination flasks.
- Excellent resiliency
- Very pliable stoppers
- Stoppers come with an 8mm-diameter hole
Stoppers have low free-sulfur content (1/2%), lower density than neoprene, and excellent resiliency and pliability. Both sizes come with an 8mm diameter hole.
Frequently Bought Together