ContactUsLinkComponent
PYREX® Hollow [ST] Reagent Bottle Stoppers, Corning
PYREX® Hollow [ST] Reagent Bottle Stoppers, Corning
Catalog # 89091-608
Supplier:  Corning
Product available on GSA Advantage®
   Low Manufacturing Impact
undefined
PYREX® Hollow [ST] Reagent Bottle Stoppers, Corning
Catalog # 89091-608
Supplier:  Corning
Supplier Number:  7575-24
Product available on GSA Advantage®
   Low Manufacturing Impact
Specifications
  • [ST] unión No.:
    24/30
  • Longitud:
    30 mm (1³⁄₁₆"), ground zone
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Certificación ambiental:
    I-RECs, ISO 14001 - Monterrey
  • Referencia:
    89091-608
  • Altura total:
    66 mm (2⅝")
  • Height Above Ground Joint:
    41 mm (1⅝")
  • Nº de proveedor:
    7575-24
Specifications
About this item
Combination glass stoppers with closed bottoms may be used as [ST] joint stoppers and as reagent bottle stoppers
They are lightweight, but strong enough to keep chipping and breakage to a minimum. Barrel-shaped head is easy to clean and rotate, side grooves prevent slippage, and flat top allows stopper to stand on head.