Accessories for PIG® Spill Kit in High-Visibility Cart, New Pig
Catálogo # 89073-968
Proveedor: 2 D 2 S.A. DE C.V. MX
Especificaciones
- Para ser utilizado con:Oils, Coolants, Solvents, Water – Universal.
- Descripción:Refill for PIG® Spill Kit in High-Visibility Cart
- Referencia:89073-968
- Absorción:53 L (14 gal.)
- Peso:6.8 kg (15 lbs.)
- Nº de proveedor:RFL244
Especificaciones
Acerca de este artículo
Don't be caught with an empty spill kit when you need it most! Keep a refill handy to instantly replace used contents in your high-visibility cart.
- Packed to refill your PIG Spill Caddy
- PIG Blue Socks stop spreading spills; PIG Rip-&-Fit Mat and Pillows quickly absorb; PIG Repair Putty plugs or patches leaks and cracks
- Temporary disposal bags with ties help make cleanup faster
- Includes gloves and goggles to guard against splashes
Only PIG Spill Kit Refills feature PIG Absorbents proven for rapid response.