Laboratory Hypodermic Needles, Cadence Science®
About this item
Cadence Science® hypodermic needles are precision made and designed to ensure strict adherence with U.S. Government specification GG-N-196. Points are machine ground to achieve uniform sharpness. Precision grinding, micro-bead jet blasting, and ultrasonic cleaning assure freedom from burrs and debris. All needle cannulae are fabricated from type 304 stainless steel tubing which must pass our rigorous in-plant quality control prior to acceptance and use.
Nonsterile. Packaged 12 needles per box.
Government specification GG-N-196. Points are machine ground to achieve uniform sharpness. Precision grinding, micro-bead jet blasting, and ultrasonic cleaning assure freedom from burrs and debris. All needle cannulae are fabricated from type 304 stainless steel tubing which must pass our rigorous in-plant quality control prior to acceptance and use.
Certifications: Plated brass luer hubs comply with luer taper specifications ISO 594.
2 Options Available Below
Product Details & Documents
Cadence Science® hypodermic needles are precision made and designed to ensure strict adherence with U.S. Government specification GG-N-196. Points are machine ground to achieve uniform sharpness. Precision grinding, micro-bead jet blasting, and ultrasonic cleaning assure freedom from burrs and debris. All needle cannulae are fabricated from type 304 stainless steel tubing which must pass our rigorous in-plant quality control prior to acceptance and use.
Nonsterile. Packaged 12 needles per box.
Government specification GG-N-196. Points are machine ground to achieve uniform sharpness. Precision grinding, micro-bead jet blasting, and ultrasonic cleaning assure freedom from burrs and debris. All needle cannulae are fabricated from type 304 stainless steel tubing which must pass our rigorous in-plant quality control prior to acceptance and use.
Certifications: Plated brass luer hubs comply with luer taper specifications ISO 594.
Documents
Detergents Cleaners, Decon.pdf
Additional Product Details
Product Family Options
Product Information
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.