ContactUsLinkComponent
SP Bel-Art Safe-Lab™ Thermometer Joint Adapters for Tapered Joints, Bel-Art Products, a part of SP
SP Bel-Art Safe-Lab™ Thermometer Joint Adapters for Tapered Joints, Bel-Art Products, a part of SP
Catalog # 70680-392
undefined
SP Bel-Art Safe-Lab™ Thermometer Joint Adapters for Tapered Joints, Bel-Art Products, a part of SP
Catalog # 70680-392
Supplier Number:  F20309-1716
Specifications
  • [ST] unión No.:
    24/40
  • Referencia:
    70680-392
  • Nº de proveedor:
    203091716
Specifications
About this item
Teflon® PTFE body with Viton® O-Ring for airtight vacuum work.
  • PTFE body construction
  • Threaded polypropylene screw cap
  • Not autoclavable
  • Designed for use with tapered joints
The adapter has a threaded polypropylene screw cap on top with an inside O-Ring for depth adjustment of 6–7 mm for straight stem thermometers, glass tubing, and other items. Rotating the extracting nut removes the adapter from the flask. The size 10/18 adapter does not have a body O-Ring. Not autoclavable.