KIMAX® Glass Pennyhead [ST] Stoppers, Flask Length, Kimble Chase
Catálogo # 89002-174
Proveedor: DWK LIFE SCIENCES S. DE R.L. DE MX
Especificaciones
- Descripción:Solid stopper
- Referencia:89002-174
- [ST] No. de tapón:13
- Nº de proveedor:41900R-13
Especificaciones
Acerca de este artículo
Stoppers are for use with flasks, mixing cylinders, separatory funnels, and other glassware
Designed from ASTM Specification E675.
Size 13 stopper is solid. Size 38 stopper is hollow.