Orders LinkContactUsLinkComponent
Falcon® Cell Strainers, Sterile, Corning®
Falcon® Cell Strainers, Sterile, Corning®
Supplier:   Corning
Falcon® Cell Strainers, Sterile, Corning®
Supplier:  Corning
Certificates
Falcon® Cell Strainers, Sterile, Corning®
Falcon® Cell Strainers, Sterile, Corning®
2 Options Available Below
Product Details & Documents
Ideal for preparing specimens for flow cytometry, these cell strainers offer a simple, ready-to-use method for obtaining uniform, single-cell suspensions. Cell strainers feature a color-coded polypropylene frame with extended tab for handling with forceps. Set within the frame is a strong nylon mesh with evenly spaced pores. Sterilized via gamma irradiation.
  • Moulded, color-coded PP frame with tab enables easy handling
  • Fits perfectly into a 50 ml Falcon® conical tube or other similarly sized tube
  • Sterilised by gamma irradiation and conveniently available in individual, peel-open packaging
  • Extended lip on strainer enables aseptic handling with forceps
Ordering information: Packaged individually in bags.
Product Family Options
Información del producto
  • Pore size
    Color
    Availability
    Price
  • 40 µm
    Blue
    Log in Para disponibilidad
    Log in Para disponibilidad
    Case (50UN)
    Case (50UN) - $7,195.94
    $7,195.94
    Case (50UN)
       $7,195.94
    Supplier Number: 352340
  • 100 µm
    Yellow
    Log in Para disponibilidad
    Log in Para disponibilidad
    Case (50UN)
    Case (50UN) - $7,089.60
    $7,089.60
    Case (50UN)
       $7,089.60
    Supplier Number: 352360