Extreme Temperature Polyimide Film Tape, Micronova
Catalog # 82030-960
Supplier: MICRONOVA MFG INC
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Specifications
- Anchura:25.4 mm (1")
- Color:Transparent amber
- Longitud:33 m (36 yds.)
- Referencia:82030-960
- Nº de proveedor:825-1AM
Specifications
About this item
This high temperature tape is constructed of transparent amber polyimide film with pressure sensitive silicone adhesive on a plastic core
The silicone adhesive ensures a clean release from most substrates, even after exposure to high temperatures. Suitable for high heat and subzero applications such as electrical insulation, lamination and test applications, subzero storage, and over-labeling. Temperature range: –73 to 260°C (–100 to 500°F).